Learn More
Fritsch Milling And Sizing Inc Grinding Bowl Cpl. for 6 Tubes Stainless Steel P-7 Pl
SDP
Catalog No. 501534632
Encompass
The grinding bowl for 6 tubes made of stainless steel for P-7 pl made of stainless steel with 6 sample glasses 1.5 ml volume is suitable for single-use technology, for example in pharmacy, forensics and pathology, RNA/DNA extraction, bacteriology, chemistry.
Additionally, a set of sample glasses 1.5 ml volume with lids (set = 12 pieces) is available with order no. 83.3165.00.
Catalog No. 50-153-4632 Supplier Fritsch Milling And Sizing Inc Supplier No. 50.9506.00
The grinding bowl for 6 tubes made of stainless steel for P-7 pl made of stainless steel with 6 sample glasses 1.5 ml volume is suitable for single-use technology, for example in pharmacy, forensics and pathology, RNA/DNA extraction, bacteriology, chemistry.
Additionally, a set of sample glasses 1.5 ml volume with lids (set = 12 pieces) is available with order no. 83.3165.00.