Learn More
Fritsch Milling And Sizing Inc Grinding Set P-2 Hardmetal Tungsten Carbide
SDP
Catalog No. 501534522
Encompass
Consisting of a high centralised mortar bowl made of hardmetal tungsten carbide rimmed in a plastic frame, pestle and scraper.
General material specification:
Material: hardmetal tungsten carbide - WC
Abrasion resistance: very good
Suitable for: hard, medium-hard, abrasive samples
Catalog No. 50-153-4522 Supplier Fritsch Milling And Sizing Inc Supplier No. 46.2080.00
Add to Cart
Add to Cart
Request a Quote
Consisting of a high centralised mortar bowl made of hardmetal tungsten carbide rimmed in a plastic frame, pestle and scraper.
General material specification:
Material: hardmetal tungsten carbide - WC
Abrasion resistance: very good
Suitable for: hard, medium-hard, abrasive samples