Learn More
Fritsch Milling And Sizing Inc Grinding Vessel Makrolon (PC)
SDP
Catalog No. 501533919
Encompass
Grinding vessels, 1,4 litre made of autoclavable, transparent and scratch-resistant plastic PC for PULVERISETTE 11.
Catalog No. 50-153-3919 Supplier Fritsch Milling And Sizing Inc Supplier No. 11.3151.16
Grinding vessels, 1,4 litre made of autoclavable, transparent and scratch-resistant plastic PC for PULVERISETTE 11.