Learn More
Electron Microscopy Sciences Potter-Elvehjem Tissue Grinder PTFE with Radial Serrations, 10 ml, 16 mmx 74 mm Mortor, 219 mm OAL
SDP
Catalog No. 501900324
Encompass_Preferred
Potter-Elvehjem Tissue Grinder incorporates a serrated pestle design to disperse homogenate into the mortar cylinder. Standard unit with Teflon pestles are well suited for soft tissue, such as brain and liver.
The tissue grinder are precisely constructed with completely interchangeable parts to allow easier cleaning or replacement of damaged mortars. This unit is autoclavable and can be used for power homogenization as with overhead stirrer. Depth of the serration is 0.20" to 0.25" (0.1 to 0.15 mm).
Size: 10 ml
Mortor: 16 mmx 74 mm
OAL: 219 mm
Catalog No. 50-190-0324 Supplier Electron Microscopy Sciences Supplier No. 6479510
Potter-Elvehjem Tissue Grinder incorporates a serrated pestle design to disperse homogenate into the mortar cylinder. Standard unit with Teflon pestles are well suited for soft tissue, such as brain and liver.
The tissue grinder are precisely constructed with completely interchangeable parts to allow easier cleaning or replacement of damaged mortars. This unit is autoclavable and can be used for power homogenization as with overhead stirrer. Depth of the serration is 0.20" to 0.25" (0.1 to 0.15 mm).
Size: 10 ml
Mortor: 16 mmx 74 mm
OAL: 219 mm