Learn More
Electron Microscopy Sciences Potter-Elvehjem Tissue Grinder PTFE Radial Serrations, 55 ml, 30 mmx 130 mm Mortor, 266 mm OAL
SDP
Catalog No. 501900327
Encompass_Preferred
Potter-Elvehjem Tissue Grinder incorporates a serrated pestle design to disperse homogenate into the mortar cylinder. Standard unit with Teflon pestles are well suited for soft tissue, such as brain and liver.
The tissue grinder are precisely constructed with completely interchangeable parts to allow easier cleaning or replacement of damaged mortars. This unit is autoclavable and can be used for power homogenization as with overhead stirrer. Depth of the serration is 0.20" to 0.25" (0.1 to 0.15 mm).
Size: 55 ml
Mortor: 30 mmx 130 mm
OAL: 266 mm
Catalog No. 50-190-0327 Supplier Electron Microscopy Sciences Supplier No. 6479555
Potter-Elvehjem Tissue Grinder incorporates a serrated pestle design to disperse homogenate into the mortar cylinder. Standard unit with Teflon pestles are well suited for soft tissue, such as brain and liver.
The tissue grinder are precisely constructed with completely interchangeable parts to allow easier cleaning or replacement of damaged mortars. This unit is autoclavable and can be used for power homogenization as with overhead stirrer. Depth of the serration is 0.20" to 0.25" (0.1 to 0.15 mm).
Size: 55 ml
Mortor: 30 mmx 130 mm
OAL: 266 mm