Learn More
Aviva Systems Biology Corp GRINL1A Antibody - C-terminal region (ARP62221_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP62221_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against GRINL1A. It was validated on Western Blot. . Alternate names: DKFZp586F1918, GCOM1, Gdown, Gdown1, GRINL1AApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Human, Mouse, Rabbit, Rat
Catalog No. 50-287-616