Learn More
Cole-Parmer Oakton Grip-Clip Probe Holder for Portable Meters
Catalog No. 501961763 Shop All Cole-Parmer Products
Encompass_Preferred
Simply clip holder on side of beaker or container to support an ATC probe and a 11-/12-mm or 16-mm probe. Allows for easy, hand-free measurement.
Catalog No. 50-196-1763 Supplier Cole-Parmer Supplier No. 3561450
Simply clip holder on side of beaker or container to support an ATC probe and a 11-/12-mm or 16-mm probe. Allows for easy, hand-free measurement.