Learn More
Checkers Grip Guard™ 3 Channel PGG3 Checkers Cable Protector
Catalog No. 19088452 Shop All Justrite Safety Group Products
Change view
Click to view available options
Color:
Yellow, Gray
Catalog No. Color
19-088-452 Yellow, Gray
1 options
Catalog No. 19-088-452 Supplier Checkers Supplier No. GG3X225Y/GRY
Only null left
Add to Cart
Add to Cart
The Grip Guard 3-Channel Cable Protector is constructed of durable, high-impact, non-conductive polyurethane, and is resistant to oils, fuels, and lubricants.
  • Grip Guard™ series 3-channel cable protectors feature Gripper Connectors with a new lightweight design
  • At a weight of only 23 lb., Grip Guard™ is up to 33% lighter than other leading polyurethane 3-channel cable protectors, and up to 50% lighter than other rubber 3-channel cable protectors on the market
  • The Gripper Connector is ergonomically designed to fit your hand, which allows for easy handling and transport
  • This connector also locks multiple cable protectors tightly together to provide a safe crossing for vehicle and pedestrian traffic while protecting valuable electrical cables and hose lines
  • When compared to other rubber manufactured cable protectors, Grip Guard™ is up to 50% lighter without compromising the required strength and durability
Specifications
Dimensions (L x W x H) 35.5 x 10.13 x 0.75 in.
Color Yellow, Gray
Material Polyurethane
Product Type Cable Protector