Learn More
MSC Albion Caster Grip Lock Brake Kit, Face Contact, 3.5 in., 1 ea.
Catalog No. 5000510683
Encompass
The grip lock brake kit provides a reliable solution for securing caster movement in material handling applications. Designed for use with 3.5 in. wheels, this accessory ensures stability during operation.
  • Face contact, pedal trailing brake type
  • Compatible with 3.5 in. wheels
  • Designed for use with series 02 casters
Catalog No. 50-005-10683 Supplier MSC Supplier No. 01859016
The grip lock brake kit provides a reliable solution for securing caster movement in material handling applications. Designed for use with 3.5 in. wheels, this accessory ensures stability during operation.
  • Face contact, pedal trailing brake type
  • Compatible with 3.5 in. wheels
  • Designed for use with series 02 casters