Learn More
Electron Microscopy Sciences Rubis Non-slip Grip Tweezers Style SS, 135 mm OAL
SDP
Catalog No. 5024077
Encompass_Preferred
This special soft coating provides a non-slip grip and improves the handling of the tweezer. The soft-coating prevents hand fatigue, and protects the hand from heat and cold. Made from the finest stainless steel grade
OAL: 135 mm
Catalog No. 50-240-77 Supplier Electron Microscopy Sciences Supplier No. 78144SS
This special soft coating provides a non-slip grip and improves the handling of the tweezer. The soft-coating prevents hand fatigue, and protects the hand from heat and cold. Made from the finest stainless steel grade
OAL: 135 mm