Learn More
Aviva Systems Biology Corp GRIP2 Antibody - C-terminal region (ARP64659_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP64659_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against GRIP2. It was validated on Western Blot. The immunogen is a synthetic peptide directed towards the C-terminal region of Human GRIP2. Alternate names: -ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Pig, Rabbit, Rat, Zebrafish
Catalog No. 50-288-327