Learn More
Aviva Systems Biology Corp GRIPAP1 Antibody - N-terminal region (ARP57358_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP57358_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against GRIPAP1. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human GRIPAP1. Alternate names: DKFZp434P0630, GRASP-1, KIAA1167, MGC126593, MGC126595ApplicationWB. Species reactivity: Cow, Human, Pig
Catalog No. 50-014-53933