Learn More
MSC PIP Grippaz Nitrile Gloves, 3XL, 7 Mil, Orange, 50/Pk.
Catalog No. 5000684476
Encompass
These 3XL, 7-mil thick nitrile gloves feature patented Grippaz™ technology with a fish scale pattern on both the inside and outside surfaces to provide superior traction when handling oily, greasy, or wet parts. Designed for durability and dexterity, these powder-free, ambidextrous gloves offer enhanced flexibility and comfort to reduce hand fatigue during prolonged use.
  • Patented fish scale texture provides unparalleled grip performance
  • More tear- and puncture-resistant than conventional disposable gloves
  • Orange color enhances eye-to-hand visual acuity
  • ESD safe per ASTM D257 (108 ohms)
  • Touchscreen compatible
  • Latex-free and silicone-free construction
  • Complies with 21 CFR, Part 177 food handling requirements
  • Ideal for automotive service, painting, printing, and equipment maintenance
Catalog No. 50-006-84476 Supplier MSC Supplier No. 67259036
These 3XL, 7-mil thick nitrile gloves feature patented Grippaz™ technology with a fish scale pattern on both the inside and outside surfaces to provide superior traction when handling oily, greasy, or wet parts. Designed for durability and dexterity, these powder-free, ambidextrous gloves offer enhanced flexibility and comfort to reduce hand fatigue during prolonged use.
  • Patented fish scale texture provides unparalleled grip performance
  • More tear- and puncture-resistant than conventional disposable gloves
  • Orange color enhances eye-to-hand visual acuity
  • ESD safe per ASTM D257 (108 ohms)
  • Touchscreen compatible
  • Latex-free and silicone-free construction
  • Complies with 21 CFR, Part 177 food handling requirements
  • Ideal for automotive service, painting, printing, and equipment maintenance