Learn More
Cole-Parmer Gripper Ferrule, 1/4"-28 UNF flat bottom, 1/16" OD tubing; 10/pk
Catalog No. NC1840115 Shop All Cole-Parmer Products
Encompass_Preferred
Available in PTFE/SS or PTFE/PEEK material.
Catalog No. NC1840115 Supplier Cole-Parmer Supplier No. 2194000
Available in PTFE/SS or PTFE/PEEK material.