Learn More
Sarstedt Inc GRIPPER RACK 20 PLACE SCMTUBES
Supplier:  Sarstedt Inc 93.848.100
Encompass_Preferred
Gripper Rack 20 Place for Screw Cap Microtubes; Polycarbonate; 10.5mm; Suitable for micro tubes with screw cap; notched aperature enables single-handed operation
Catalog No. NC9107002