Learn More
Curiox Biosystems Inc GRIPPER/TILTER PACK
SDP
Catalog No. NC3650102
Encompass
  • Designed for seamless integration with the C-FREE Pluto System.
  • Enhances automated liquid handling capabilities.
  • Precisely grips microplates for efficient processing.
  • Facilitates plate movement for tilting operations.
  • Reduces manual intervention in scientific workflows.
  • Supports consistent cell sample preparation.
  • Available as a package with the Pluto System Tilter or as a replacement.
  • Integrates with Pluto wash and laminar mix technologies.
  • Applicable in antibody cocktailing, cell staining, and cell washing.
  • Reduces manual effort and potential for human error in workflows.
Catalog No. NC3650102 Supplier Curiox Biosystems Inc Supplier No. CFPLUGRT01
Add to Cart
Add to Cart
Request a Quote
  • Designed for seamless integration with the C-FREE Pluto System.
  • Enhances automated liquid handling capabilities.
  • Precisely grips microplates for efficient processing.
  • Facilitates plate movement for tilting operations.
  • Reduces manual intervention in scientific workflows.
  • Supports consistent cell sample preparation.
  • Available as a package with the Pluto System Tilter or as a replacement.
  • Integrates with Pluto wash and laminar mix technologies.
  • Applicable in antibody cocktailing, cell staining, and cell washing.
  • Reduces manual effort and potential for human error in workflows.