Learn More
Electron Microscopy Sciences Gripper Tool for Cryo Grid Boxes
SDP
Catalog No. 501929973
Encompass_Preferred
Use this tool to open the lid of our cryogrid box (EMS Cat. #71166-40). It features a spring loaded vise that grips the lid securely and a ribbed barrel for ease in unscrewing the lid. Made of high quality aluminum, it can be used with liquid nitrogen. Can also be used to carry the cryo grid box.
Catalog No. 50-192-9973 Supplier Electron Microscopy Sciences Supplier No. 71166SP
Use this tool to open the lid of our cryogrid box (EMS Cat. #71166-40). It features a spring loaded vise that grips the lid securely and a ribbed barrel for ease in unscrewing the lid. Made of high quality aluminum, it can be used with liquid nitrogen. Can also be used to carry the cryo grid box.