Learn More
Mitegen Metal Cryo Grid Box Gripper Tool For SKU M-CEM-AGG1001, Aluminum, For Secure Transfer of Metal Cryo Grid Boxes with Pin-Type Lids
SDP
Catalog No. NC1816923
Encompass
This Metal Cryo Grid Box Gripper Tool is specifically designed for the secure and efficient transfer of various grid boxes, particularly the Metal Cryo Grid Box with a pin-type lid (SKU M-CEM-AGG1001). Crafted from durable aluminum, this gripper ensures a firm, stable hold on grid boxes, minimizing direct hand contact and reducing the risk of accidental drops or thermal shock to delicate cryogenic samples during transfer between locations. Its robust construction provides enhanced longevity and reliability in demanding laboratory conditions. This tool is ideally suited for moving grid boxes between dewars, workstations, or into automated systems. It is intended solely for transferring grid boxes and should not be used to open or unscrew lids, ensuring proper handling procedures are maintained.
Catalog No. NC1816923 Supplier Mitegen Supplier No. MCEMAGT1003
This Metal Cryo Grid Box Gripper Tool is specifically designed for the secure and efficient transfer of various grid boxes, particularly the Metal Cryo Grid Box with a pin-type lid (SKU M-CEM-AGG1001). Crafted from durable aluminum, this gripper ensures a firm, stable hold on grid boxes, minimizing direct hand contact and reducing the risk of accidental drops or thermal shock to delicate cryogenic samples during transfer between locations. Its robust construction provides enhanced longevity and reliability in demanding laboratory conditions. This tool is ideally suited for moving grid boxes between dewars, workstations, or into automated systems. It is intended solely for transferring grid boxes and should not be used to open or unscrew lids, ensuring proper handling procedures are maintained.