Learn More
Medchemexpress LLC Griseofulvin-13C,d3 | 1329612-29-4 | 356.78 | 1 MG
SDP
Catalog No. 5000387115
Encompass_Preferred
Griseofulvin-13C,d3 is the 13C- and deuterium labeled Griseofulvin. It is for research use only and not sold to patients.
  • Can be used as a tracer.
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
Catalog No. 50-003-87115 Supplier Medchemexpress LLC Supplier No. HY17583S11MG
Griseofulvin-13C,d3 is the 13C- and deuterium labeled Griseofulvin. It is for research use only and not sold to patients.
  • Can be used as a tracer.
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.