Learn More
Cayman Chemical GrIseofulvIn-d3 1mg
SDP
Supplier:  Cayman Chemical 333661 MG
Encompass_Preferred
An internal standard for the quantification of griseofulvin by GC- or LC-MS
Catalog No. 50-294-8958