Learn More
Medchemexpress LLC Griseofulvin (standard) | 126-07-8 | 98.9% | 352.77 | 500 MG
SDP
Catalog No. 5000387503
Encompass_Preferred
Griseofulvin is an orally active antifungal antibiotic with antitumor activity. It induces apoptosis and G2/M cell cycle arrest in cancer cells. Griseofulvin also possesses cardiovascular modulatory activity, which includes reducing angina pectoris, relieving hand artery spasm associated with onychomycosis, and treating peripheral vascular diseases such as shoulder-hand syndrome.
  • Orally active antifungal antibiotic
  • Antitumor activity
  • Induces apoptosis and G2/M cell cycle arrest in cancer cells
  • Cardiovascular modulatory activity
  • Reduces angina pectoris
  • Relieves hand artery spasm associated with onychomycosis
  • Treats peripheral vascular diseases like shoulder-hand syndrome
Catalog No. 50-003-87503 Supplier Medchemexpress LLC Supplier No. HY17583500MG
Griseofulvin is an orally active antifungal antibiotic with antitumor activity. It induces apoptosis and G2/M cell cycle arrest in cancer cells. Griseofulvin also possesses cardiovascular modulatory activity, which includes reducing angina pectoris, relieving hand artery spasm associated with onychomycosis, and treating peripheral vascular diseases such as shoulder-hand syndrome.
  • Orally active antifungal antibiotic
  • Antitumor activity
  • Induces apoptosis and G2/M cell cycle arrest in cancer cells
  • Cardiovascular modulatory activity
  • Reduces angina pectoris
  • Relieves hand artery spasm associated with onychomycosis
  • Treats peripheral vascular diseases like shoulder-hand syndrome