Learn More
Eurofins Discoverx GRK1, active, 10 µg. (14-935)
Supplier:  Eurofins Discoverx 14935
Encompass
N-terminal 6xHis-tagged recombinant, human full length GRK1. Retina-specific kinase involved in the signal turnoff via phosphorylation of rhodopsin (RHO), the G protein- coupled receptor that initiates the phototransduction cascade. This rapid desensitization is essential for scotopic vision and permits rapid adaptation to changes in illumination. For detailed product information, visit www.discoverx.com/proteins.
Catalog No. 50-199-9443