Learn More
Aviva Systems Biology Corp GRK4 Antibody - middle region (ARP53566_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP53566_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against GRK4. It was validated on Western Blot. The immunogen is a synthetic peptide directed towards the middle region of human GRK4. Alternate names: GPRK2L, GPRK4, GRK4a, IT11ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rat, Zebrafish
Catalog No. 50-285-850