Learn More
Selleck Chemical LLC GRK5-IN-2 E2817-25MG
SDP
Catalog No. 5000040027
Encompass_Preferred
GRK5-IN-2 is a potent G-protein-coupled receptor kinase 5 (GRK5) inhibitor
Catalog No. 50-000-40027 Supplier Selleck Chemical LLC Supplier No. E281725MG
GRK5-IN-2 is a potent G-protein-coupled receptor kinase 5 (GRK5) inhibitor