Learn More
Medchemexpress LLC HY-136561 5mg , GRK5-IN-2 CAS:1642839-27-7 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY1365615MG
Encompass_Preferred
HY-136561 5mg GRK5-IN-2 CAS: 1642839-27-7 GRK5-IN-2 (compound 707), a pyridine-based bicyclic compound, is a potent G-protein-coupled receptor kinase 5 (GRK5) inhibitor. GRK5-IN-2 regulates the expression and/or release of insulin and is useful for the metabolic disease research. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-226-1408