Learn More
Aviva Systems Biology Corp GRK7 Antibody - C-terminal region (ARP59819_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP59819_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against GRK7. It was validated on Western Blot. The immunogen is a synthetic peptide directed towards the C terminal region of human GRK7. Alternate names: GPRK7ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Rat
Catalog No. 50-286-842