Learn More
Medchemexpress LLC GRL-0496 | 1087243-14-8 | 99.2% | 272.69 | 100 MG
SDP
Supplier:  Medchemexpress LLC HY137954100MG
Encompass_Preferred
GRL-0496 is a potent chloropyridyl ester-derived SARS-CoV 3CLpro inhibitor. It demonstrates an IC50 of 30 nM in both enzyme inhibitory and antiviral assays. It also exhibits SARS-CoV antiviral activity with an EC50 of 6.9 μM.
  • Potent chloropyridyl ester-derived SARS-CoV 3CLpro inhibitor
  • IC50 of 30 nM in enzyme inhibitory assays
  • IC50 of 30 nM in antiviral assays
  • SARS-CoV antiviral activity with an EC50 of 6.9 μM
Catalog No. 50-003-51359