Learn More
Medchemexpress LLC GRL-0496 | 1087243-14-8 | 99.20% | 272.69 | 50 MG
SDP
Catalog No. 5000351613
Encompass_Preferred
GRL-0496 is a potent chloropyridyl ester-derived SARS-CoV 3CLpro inhibitor, demonstrating an IC50 of 30 nM in both enzyme inhibitory and antiviral assays. It exhibits SARS-CoV antiviral activity with an EC50 of 6.9 μM.
  • Potent chloropyridyl ester-derived inhibitor
  • Exhibits SARS-CoV antiviral activity
  • Intended for research use only
Catalog No. 50-003-51613 Supplier Medchemexpress LLC Supplier No. HY13795450MG
GRL-0496 is a potent chloropyridyl ester-derived SARS-CoV 3CLpro inhibitor, demonstrating an IC50 of 30 nM in both enzyme inhibitory and antiviral assays. It exhibits SARS-CoV antiviral activity with an EC50 of 6.9 μM.
  • Potent chloropyridyl ester-derived inhibitor
  • Exhibits SARS-CoV antiviral activity
  • Intended for research use only