Learn More
Medchemexpress LLC GRL-1720 TFA | 2835511-03-8 | C16H12ClF3N2O4 | 5 MG
SDP
Catalog No. 5000372716
Encompass_Preferred
GRL-1720 TFA is a potent inhibitor of SARS-CoV-2 Mpro, exhibiting anti-SARS-CoV2 activity with an EC50 value of 15 μM. This compound has a molecular weight of 388.73 and appears as a solid, ranging from light brown to brown in color.
  • High purity of 98.94%.
  • Shipped at room temperature.
  • Store at -20°C, sealed, away from moisture.
  • In solution, stable for 6 months at -80°C or 1 month at -20°C with sealed storage, away from moisture.
Catalog No. 50-003-72716 Supplier Medchemexpress LLC Supplier No. HY153533A5MG
GRL-1720 TFA is a potent inhibitor of SARS-CoV-2 Mpro, exhibiting anti-SARS-CoV2 activity with an EC50 value of 15 μM. This compound has a molecular weight of 388.73 and appears as a solid, ranging from light brown to brown in color.
  • High purity of 98.94%.
  • Shipped at room temperature.
  • Store at -20°C, sealed, away from moisture.
  • In solution, stable for 6 months at -80°C or 1 month at -20°C with sealed storage, away from moisture.