Learn More
Medchemexpress LLC HY-117043 10mg Medchemexpress, GRL0617 CAS:1093070-16-6 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY11704310MG
Encompass_Preferred
Medchemexpress, HY-117043 10mg GRL0617 CAS:1093070-16-6 GRL0617 is a potent, selective and competitive noncovalent inhibitor of severe acute respiratory syndrome (SARS-CoV) papain-like protease (PLpro)/deubiquitinase, with an IC50 of 0.6 μM, and with a Ki of 0.49 μM. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-196-9448