Learn More
Newcomer Supply Gram, Hucker-Twort Stain Kit, 250 ml kit
SDP
Catalog No. NC9119886
Encompass
Newcomer Supply Gram, Hucker-Twort Stain Kit is a simple, rapid procedure for staining gram-positive and gram-negative bacteria without picric acid.  The Twort Stain combines Neutral Red and Fast Green, for clear detection of red gram-negative bacteria with a green counterstain. Shelf Life is 2 years from date of manufacture. Store at 15-30°C.
Catalog No. NC9119886 Supplier Newcomer Supply Supplier No. 9125A
Newcomer Supply Gram, Hucker-Twort Stain Kit is a simple, rapid procedure for staining gram-positive and gram-negative bacteria without picric acid.  The Twort Stain combines Neutral Red and Fast Green, for clear detection of red gram-negative bacteria with a green counterstain. Shelf Life is 2 years from date of manufacture. Store at 15-30°C.