Learn More
Aviva Systems Biology Corp GRM6 Antibody - C-terminal region (ARP32597_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP32597_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against GRM6. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the C terminal region of human GRM6. Alternate names: mGlu6, CSNB1B, GPRC1F, MGLUR6ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Yeast, Zebrafish
Catalog No. 50-014-54669