Learn More
Aviva Systems Biology Corp GRM6 Antibody - N-terminal region (ARP58004_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP58004_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against GRM6. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human GRM6. Alternate names: CSNB1B, DKFZp686H1993, GPRC1F, MGLUR6, mGlu6ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Pig, Rabbit, Rat, Sheep, Zebrafish
Catalog No. 50-014-62209