Learn More
Aviva Systems Biology Corp GRM7 Antibody - C-terminal region (ARP59604_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP59604_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against GRM7. It was validated on Western Blot. The immunogen is a synthetic peptide directed towards the C terminal region of human GRM7. Alternate names: FLJ40498, GLUR7, GPRC1G, MGLU7, MGLUR7ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Zebrafish
Catalog No. 50-286-754