Learn More
Bellco Glass, Inc. Grn Open top S/Cap,45mm With 1 5/16 Hole"
SDP
Catalog No. 501945750
Encompass_Preferred
Ideal for addition & withdraw of contents via syringe. 45mm cap has 33mm hole to accommodate 45mm septa or 45mm port assembly. 32mm cap has 20mm hole to accommodate 32mm septa or 32mm port assembly. MADE IN THE USA Polypropylene Size: 45mm Color: green
Catalog No. 50-194-5750 Supplier Bellco Glass, Inc. Supplier No. 563700015
Ideal for addition & withdraw of contents via syringe. 45mm cap has 33mm hole to accommodate 45mm septa or 45mm port assembly. 32mm cap has 20mm hole to accommodate 32mm septa or 32mm port assembly. MADE IN THE USA Polypropylene Size: 45mm Color: green