Learn More
Electron Microscopy Sciences Green Pestle And 1.5 ml Tube
SDP
Catalog No. 501899195
Encompass_Preferred
Disposable polyethylene pestles and 1.5 ml micro centrifuge tubes are used to resuspend precipitates such as protein of DNA pellets in buffer solution or to grind soft tissue. The pestles are in a matched fit for the tubes. These sets are excellent for processing large numbers of samples.
Catalog No. 50-189-9195 Supplier Electron Microscopy Sciences Supplier No. 6478810
Disposable polyethylene pestles and 1.5 ml micro centrifuge tubes are used to resuspend precipitates such as protein of DNA pellets in buffer solution or to grind soft tissue. The pestles are in a matched fit for the tubes. These sets are excellent for processing large numbers of samples.