Learn More
Cayman Chemical Granisetron-d3, Purity: ≥99% deuterated forms (d1-d3), CAS: 1224925-76-1, 1 mg
SDP
Catalog No. 502992603
Encompass_Preferred
An internal standard for the quantification of granisetron by GC- or LC-MS
Catalog No. 50-299-2603 Supplier Cayman Chemical Supplier No. 317861 MG
An internal standard for the quantification of granisetron by GC- or LC-MS