Learn More
Cayman Chemical Grnzyme B MoncLNal ntIbod 50uL
SDP
Catalog No. 502915161
Encompass_Preferred
An a-glucosidase inhibitor (IC50 0.7 Ug/ml) and a prodrug form of castanospermine reduces BVDV levels in a plaque formation assay (IC50 16 mM) and increases survival of MDBK infected with BVDV (IC50 47 UM) reduces viremia and increases survival of mice infected with a lethal dose of dengue virus at 50 mg/kg twice daily i.p.
Catalog No. 50-291-5161 Supplier Cayman Chemical Supplier No. 3706750 UL
An a-glucosidase inhibitor (IC50 0.7 Ug/ml) and a prodrug form of castanospermine reduces BVDV levels in a plaque formation assay (IC50 16 mM) and increases survival of MDBK infected with BVDV (IC50 47 UM) reduces viremia and increases survival of mice infected with a lethal dose of dengue virus at 50 mg/kg twice daily i.p.