Learn More
Cayman Chemical Grnzyme BCTerm RabbIt Mo 100uL
SDP
Catalog No. 502911245
Encompass_Preferred
An androgenically inactive metabolite of testosterone and androstenedione potentiates GABA-induced chloride currents in HEK293 cells expressing rat a1B2Y2L subunit-containing GABAA receptors at 10 UM protective against seizures induced by 6 Hz electroshock or PTZ (ED50s 76 9 and 139 mg/kg respectively) increases body temperature in rhesus macaques
Catalog No. 50-291-1245 Supplier Cayman Chemical Supplier No. 35806100 UL
An androgenically inactive metabolite of testosterone and androstenedione potentiates GABA-induced chloride currents in HEK293 cells expressing rat a1B2Y2L subunit-containing GABAA receptors at 10 UM protective against seizures induced by 6 Hz electroshock or PTZ (ED50s 76 9 and 139 mg/kg respectively) increases body temperature in rhesus macaques