Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC GRO-alpha/CXCL1 Hum 10ug
SDP

Catalog No. 5000185474
Encompass_Preferred

Recombinant human GRO-alpha (CXCL1) is a HEK293-expressed, His-tagged chemokine supplied as a lyophilized powder. It contains the mature sequence (residues A35-N107), is low in endotoxin, and is reported active in chemotaxis and proliferation assays. The reagent is designed for functional and biochemical studies, including receptor activation and cell signaling assays; aliquot and freeze for extended stability.

  • HEK293-expressed, His-tagged recombinant human CXCL1.
  • Contains mature sequence A35-N107.
  • Supplied as a lyophilized powder formulated with trehalose for stability.
  • Low endotoxin level (<1 EU/μg).
  • Demonstrated bioactivity with ED50 ≈ 3.5-3.7 ng/mL in functional assays.
  • Suitable for chemotaxis, receptor activation, and signaling studies.
  • Store at -20°C; aliquot and freeze at -80°C for long-term storage.

Catalog No. 50-001-85474 Supplier Medchemexpress LLC Supplier No. HYP7050810UG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Recombinant human GRO-alpha (CXCL1) is a HEK293-expressed, His-tagged chemokine supplied as a lyophilized powder. It contains the mature sequence (residues A35-N107), is low in endotoxin, and is reported active in chemotaxis and proliferation assays. The reagent is designed for functional and biochemical studies, including receptor activation and cell signaling assays; aliquot and freeze for extended stability.

  • HEK293-expressed, His-tagged recombinant human CXCL1.
  • Contains mature sequence A35-N107.
  • Supplied as a lyophilized powder formulated with trehalose for stability.
  • Low endotoxin level (<1 EU/μg).
  • Demonstrated bioactivity with ED50 ≈ 3.5-3.7 ng/mL in functional assays.
  • Suitable for chemotaxis, receptor activation, and signaling studies.
  • Store at -20°C; aliquot and freeze at -80°C for long-term storage.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.