Learn More
Medchemexpress LLC GRO-alpha/CXCL1 Mou 10ug
SDP
Catalog No. 5000182385
Encompass_Preferred
CXCL1 (Chemokine (C-X-C motif) ligand 1) also known as GRO alpha NAP-3 or MGSA belongs to the sub-family of CXC chemokine CXCL1 is involved in the development of many inflammatory diseases including the induction of angiogenesis and recruitment of neutrophils CXCL1 is produced by many cell types and activates CXCR2 and at high levels CXCR1[1] GRO-alpha/CXCL1 Protein Mouse (HEK293 His) is produced in HEK293 cells with six C-Terminal His-tags It consists of 77 amino acids (A20-N96)
Catalog No. 50-001-82385 Supplier Medchemexpress LLC Supplier No. HYP7268410UG
CXCL1 (Chemokine (C-X-C motif) ligand 1) also known as GRO alpha NAP-3 or MGSA belongs to the sub-family of CXC chemokine CXCL1 is involved in the development of many inflammatory diseases including the induction of angiogenesis and recruitment of neutrophils CXCL1 is produced by many cell types and activates CXCR2 and at high levels CXCR1[1] GRO-alpha/CXCL1 Protein Mouse (HEK293 His) is produced in HEK293 cells with six C-Terminal His-tags It consists of 77 amino acids (A20-N96)