Learn More
Medchemexpress LLC HY-P7188 10ug Medchemexpress, GRO-alpha/CXCL1, Mouse CAS: Purity:>98%
SDP
Catalog No. 501970579
Encompass_Preferred
Medchemexpress, HY-P7188 10ug GRO-alpha/CXCL1, Mouse CAS: Recombinant Mouse GRO alpha/CXCL1 could play a major role in inflammation, angiogenesis, tumorigenesis, and wound healing. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-197-0579 Supplier Medchemexpress LLC Supplier No. HYP718810UG
Medchemexpress, HY-P7188 10ug GRO-alpha/CXCL1, Mouse CAS: Recombinant Mouse GRO alpha/CXCL1 could play a major role in inflammation, angiogenesis, tumorigenesis, and wound healing. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.