Learn More
Medchemexpress LLC HY-P7190 10ug Medchemexpress, GRO-beta/CXCL2, Human CAS: Purity:>98%
SDP
Catalog No. 501970580
Encompass_Preferred
Medchemexpress, HY-P7190 10ug GRO-beta/CXCL2, Human CAS: Recombinant Human GRO beta/CXCL2 is a powerful neutrophil chemoattractant and is involved in many immune responses including wound healing, cancer metastasis, and angiogenesis. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-197-0580 Supplier Medchemexpress LLC Supplier No. HYP719010UG
Medchemexpress, HY-P7190 10ug GRO-beta/CXCL2, Human CAS: Recombinant Human GRO beta/CXCL2 is a powerful neutrophil chemoattractant and is involved in many immune responses including wound healing, cancer metastasis, and angiogenesis. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.