Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC GRO-beta/CXCL2 Rat 10ug
SDP

Catalog No. 5000196361
Encompass_Preferred

GRO-beta/CXCL2 Protein, Rat is a recombinant chemokine used to study neutrophil chemotaxis, inflammation, angiogenesis, and tumor metastasis. Supplied lyophilized and produced in E. coli, the protein corresponds to residues S32-N100 and shows high activity in CXCR2-based assays; follow the datasheet for reconstitution and storage recommendations to preserve biological activity.

  • High specific activity suitable for receptor activation and chemotaxis assays.
  • Low endotoxin level for use in sensitive biological experiments.
  • Lyophilized format for convenient storage and transport.
  • Recombinant sequence corresponds to residues S32-N100 for sequence consistency.
  • Stable when stored frozen; aliquot after reconstitution to preserve activity.
  • Reconstitution guidance recommends a minimum concentration of 100 μg/mL in ddH2O.

Catalog No. 50-001-96361 Supplier Medchemexpress LLC Supplier No. HYP719110UG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

GRO-beta/CXCL2 Protein, Rat is a recombinant chemokine used to study neutrophil chemotaxis, inflammation, angiogenesis, and tumor metastasis. Supplied lyophilized and produced in E. coli, the protein corresponds to residues S32-N100 and shows high activity in CXCR2-based assays; follow the datasheet for reconstitution and storage recommendations to preserve biological activity.

  • High specific activity suitable for receptor activation and chemotaxis assays.
  • Low endotoxin level for use in sensitive biological experiments.
  • Lyophilized format for convenient storage and transport.
  • Recombinant sequence corresponds to residues S32-N100 for sequence consistency.
  • Stable when stored frozen; aliquot after reconstitution to preserve activity.
  • Reconstitution guidance recommends a minimum concentration of 100 μg/mL in ddH2O.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.