Learn More
Medchemexpress LLC GRO-gamma/CXCL3 Hum 10ug
SDP
Catalog No. 5000196136
Encompass_Preferred
Recombinant human GRO-gamma (CXCL3) is a CXC chemokine that functions as a neutrophil chemoattractant via CXCR2. Supplied lyophilized and expressed in E. coli, the protein is validated for chemotaxis bioassays and designed for research use in studies of leukocyte chemotaxis, cell migration, angiogenesis, and CXCR2-mediated signaling pathways.
  • Approximately 7.9 kDa, 73 amino acids.
  • Expressed in E. coli and supplied tag free.
  • Purity greater than 95% by reducing SDS-PAGE.
  • Endotoxin level below 1 EU/μg (LAL method).
  • Lyophilized formulation from 20 mM PB, pH 7.4, 50 mM NaCl.
  • Active in chemotaxis bioassays at 10-100 ng/mL.
  • Stable when stored lyophilized at -20°C; aliquot and freeze for long-term storage.
  • Available in small pack sizes for assay optimization.
Catalog No. 50-001-96136 Supplier Medchemexpress LLC Supplier No. HYP719310UG
Recombinant human GRO-gamma (CXCL3) is a CXC chemokine that functions as a neutrophil chemoattractant via CXCR2. Supplied lyophilized and expressed in E. coli, the protein is validated for chemotaxis bioassays and designed for research use in studies of leukocyte chemotaxis, cell migration, angiogenesis, and CXCR2-mediated signaling pathways.
  • Approximately 7.9 kDa, 73 amino acids.
  • Expressed in E. coli and supplied tag free.
  • Purity greater than 95% by reducing SDS-PAGE.
  • Endotoxin level below 1 EU/μg (LAL method).
  • Lyophilized formulation from 20 mM PB, pH 7.4, 50 mM NaCl.
  • Active in chemotaxis bioassays at 10-100 ng/mL.
  • Stable when stored lyophilized at -20°C; aliquot and freeze for long-term storage.
  • Available in small pack sizes for assay optimization.