Learn More
Cosmo Bio Usa Inc GroE Mix Kit
SDP
Catalog No. NC1359833
Encompass
GroE Mix is a newly developed molecular chaperone supplement for use in cell-free coupled transcription/translation systems PUREfrex 2.0 and PUREfrex 2.1 to assist proper protein folding, activity and solubility. PUREfrex 2.0 and PUREfrex 2.1 contain no molecular chaperones. When proper protein folding, activity and solubility requires molecular chaperones GroE Mix could be an ideal solution. GroE Mix comprises an optimized mixture of highly purified E. coli. GroEL (Hsp60) and GroES.
Catalog No. NC1359833 Supplier Cosmo Bio Usa Inc Supplier No. GFKPF0040.5EX
GroE Mix is a newly developed molecular chaperone supplement for use in cell-free coupled transcription/translation systems PUREfrex 2.0 and PUREfrex 2.1 to assist proper protein folding, activity and solubility. PUREfrex 2.0 and PUREfrex 2.1 contain no molecular chaperones. When proper protein folding, activity and solubility requires molecular chaperones GroE Mix could be an ideal solution. GroE Mix comprises an optimized mixture of highly purified E. coli. GroEL (Hsp60) and GroES.