Learn More
Enzo Life Sciences GroEL (E. coli), (recombinant) (50µg)
SDP
Supplier:  Enzo Life Sciences ADISPP610D
Encompass_Preferred
Alternative name: Heat shock protein 60, 60kDa chaperonin, Protein Cpn60 GroEL protein, HSP60. Purity: ≥90% (SDS-PAGE; Western blot). MW: ~60kDa. Formulation: Liquid. In 50mM TRIS-HCl, pH 7.5, containing 100 mM sodium chloride, 10mM magnesium chloride, 5mM DTT, and 0.05% sodium azide. Source: Produced in E. coli. Long Term Storage: -80°C. Applications: ELISA, WB in vitro Assay, Antigen microarray, Activity assay.
Catalog No. 50-200-7610