Learn More
Enzo Life Sciences GroEL (E. coli) monoclonal antibody (9A1/2) (50 µg)
SDP
Supplier:  Enzo Life Sciences ADISPS870D
Encompass_Preferred
Clone: 9A1/2. Host: Mouse. Isotype: IgG1. Immunogen: E. coli GroEL. UniProt: P0A6F5 (strain K12). Source: Purified from mouse ascites. Species Reactivity: E. coli. Applications: IP, WB. Recommended dilutions: Western Blot (1:10,000, ECL). Suggested dilutions/conditions may not be available for all applications. Optimal conditions must be determined individually for each application. Purity: Protein G affinity purified. Formulation: Liquid. In PBS containing 50% glycerol and 0.09% sodium azide. Handling: Avoid freeze/thaw cycles.
Catalog No. 50-667-799