Learn More
Enzo Life Sciences GroEL (E. coli) polyclonal antibody (50 µg)
SDP
Supplier:  Enzo Life Sciences ADISPS875D
Encompass_Preferred
Host: Rabbit. Immunogen: E. coli GroEL. UniProt: P0A6F5 (strain K12). Source: Purified from rabbit serum. Species reactivity: E. coli. Applications: WB. Recommended dilutions: Western Blot (1:1,000, colorimetric). Suggested dilutions/conditions may not be available for all applications. Optimal conditions must be determined individually for each application. Purity: Protein A affinity purified. Formulation: Liquid. In PBS containing 50% glycerol and 0.09% sodium azide. Handling: Avoid freeze/thaw cycles.
Catalog No. 50-950-116