Learn More
Enzo Life Sciences GroEL (E. coli), (recombinant) (200 µg)
SDP
Supplier:  Enzo Life Sciences ADISPP610F
Encompass_Preferred
Alternative name: Heat shock protein 60, 60kDa chaperonin, Protein Cpn60 GroEL protein, HSP60. MW: ~60kDa. Purity: ≥90% (SDS-PAGE; Western blot). Formulation: Liquid. In 50mM TRIS-HCl, pH 7.5, containing 100 mM sodium chloride, 10mM magnesium chloride, 5mM DTT, and 0.05% sodium azide. Source: Produced in E. coli. UniProt: P0A6F5 (strain K12). Application: ELISA, WB, in vitro Assay, Antigen microarray, Activity assay. Long Term Storage: -80°C
Catalog No. 50-667-728